Learn More
Advanced ImmunoChemical, Inc. CA19-9, Human Metastatic Liver Carcinoma, 250 KIU
Supplier:  Advanced ImmunoChemical, Inc. 8CA199250KIU
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Antigens / Tumor Markers / CA19-9. Source:Human metastatic liver carcinoma.Blood sample from the tissue donor was tested and found negative for HBsAg, HIV-1 and HIV-2 antibodies, HCV, and syphilis.Applications:Immunogen, calibrator in immunoassays, in vitro studies.Specific activity:Specific activity confirmed by enzyme immunoassay.Presentation:PBS, pH 7.4, 3 % sucrose, 0.05 % sodium azide (NaN3).Storage Conditions:-20 °C (between -15 and -30 °C allowed)Material safety note:This product is sold for research use only. Standard Laboratory Practices should be followed when handling this material.Product contains sodium azide as a preservative. Although the amount of sodium azide is very small appropriate care must be taken when handling this product.
Catalog No. 50-153-9850