Learn More
Apexbio Technology LLC HyperPFU high-fidelity DNA polymerase 1000U
SDP
Supplier:  Apexbio Technology LLC K10311000
Encompass_Preferred
Phusion High-Fidelity DNA Polymerase is a fusion enzyme composed of a Pyrococcus-like proofreading polymerase and a DNA binding domain Phusion DNA Polymerase generates PCR products with superior accuracy and speed Additionally it is highly tolerant to various inhibitors allowing robust amplification with minimal optimization steps Phusion DNA Polymerase is ideally suited for cloning applications including the amplification of long or challenging (GC-rich) DNA fragments Phusion High-Fidelity DNA Polymerase is among the most accurate thermostable polymerases available displaying a greater than 50-fold lower error rate compared to Taq DNA polymerase and 6-fold lower error rate compared to Pyrococcus furiosus (Pfu) DNA polymerase Phusion DNA Polymerase possesses 5 to 3 polymerase activity and 3 to 5 exonuclease (proofreading) activity and produces blunt-end PCR products It is supplied with a standard 5X Phusion HF Buffer suitable for complex or GC-rich templates
Catalog No. 50-310-7132