Learn More
Genscript Corporation CA242 1A6 mAb Mouse
Supplier:  Genscript Corporation V06001100
Encompass
CA242 known as carbohydrate antigen 242 is a sialyated carbohydrate antigen It is a cancer-associated glycoprotein expressed in the sera of pancreatic cancer patients CA242 is a useful biomarker for the diagnosis prognosis and surveillance of gastrointestinal malignancies
Catalog No. 50-000-58551
Add to Cart
Request a Quote