Learn More
Sino Biological Assay Buffer and Co-factors: Ca2+/Calmodulin Solution II, 10x
Supplier:  Sino Biological C0239B500
Encompass_Preferred
10x stock solution used for assaying activity of calcium-calmodulin-activated protein kinase enzymes.
Catalog No. 50-256-9657