Learn More
Selleck Chemical LLC CA3
SDP
Supplier:  Selleck Chemical LLC S866125MG
Encompass_Preferred
CA3 (CIL56) has potent inhibitory effects on YAP1/Tead transcriptional activity and primarily targets YAP1 high and therapy-resistant esophageal adenocarcinoma cells endowed with CSC properties CA3(CIL56) induces ferroptosis and iron-dependent reactive oxygen species (ROS)
Catalog No. 50-000-00999