Learn More
Hach Company CA610 Fluoride Reagent Set
Supplier:  Hach Company 2816900
Encompass
CA610 Fluoride Analyzer Reagent Set includes Reagent 1 (TISAB) Standard 1 0 5 mg/L as F Standard 2 5 0 mg/L as F
Catalog No. 50-002-56940