Learn More
Selleck Chemical LLC CA77.1
SDP
Supplier:  Selleck Chemical LLC S63955MG
Encompass_Preferred
CA77 1 (CA) is a potent brain-penetrant and orally active activator of chaperone-mediated autophagy (CMA) for the treament of Alzheimer s disease (AD)
Catalog No. 50-000-00031