Learn More
Aviva Systems Biology Corp CA8 Antibody - N-terminal region (ARP42166_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42166_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CA8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CA8. Alternate names: CA-VIII, CALS, CARP, MGC120502, MGC99509, CAMRQ3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58128