Learn More
Medchemexpress LLC Cabamiquine succinate | 2444781-71-7 | 99.8% | 580.65 | C31H37FN4O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117684A100MG
Encompass_Preferred
Cabamiquine succinate is the succinate salt of cabamiquine (DDD107498), a potent orally active antimalarial that inhibits Plasmodium protein synthesis by targeting the parasite elongation factor eEF2. It exhibits low-nanomolar cellular potency and is provided for research use under controlled storage conditions.
  • Potent antimalarial activity against Plasmodium species.
  • Targets parasite elongation factor eEF2 to inhibit protein synthesis.
  • Low-nanomolar potency in cellular assays.
  • High chemical purity appropriate for research applications.
  • Stable when stored sealed away from moisture and light.
Catalog No. 50-002-26471