Learn More
Apexbio Technology LLC Cabazitaxel 183133-96-2 100mg
SDP
Supplier:  Apexbio Technology LLC B2157100
Encompass_Preferred
Cabazitaxel (CAS 183133-96-2) is a semisynthetic compound derived from the natural precursor 10-deacetylbaccatin III belonging to the taxoid class of antineoplastic agents It exerts its biological effects primarily by stabilizing microtubules thus disrupting mitotic and interphase cellular functions and causing cell-cycle arrest leading to apoptosis Due to this microtubule-targeting activity cabazitaxel is extensively studied in oncology research particularly investigating chemoresistant tumor cell lines and therapeutic approaches for taxane-resistant cancers
Catalog No. 50-000-62178