Learn More
Selleck Chemical LLC Cabazitaxel 5mg 183133-96-2 RPR-116258A, XRP6258, TXD 258
SDP
Supplier:  Selleck Chemical LLC S30225MG
Encompass_Preferred
Cabazitaxel (RPR-116258A, XRP6258, TXD 258) is a semi-synthetic derivative of a natural taxoid that kills cancer cells by inhibiting cell division and growth. Cabazitaxel exerts its effects by inhibiting microtubule growth and assembly, processes that are essential for cells to divide. Cabazitaxel induces autophagy via the PI3K/Akt/mTOR pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3266