Learn More
Medchemexpress LLC Cabazitaxel-d6 | 1383561-29-2 | 97.9% | 841.97 | 1 MG
SDP
Supplier:  Medchemexpress LLC 1MGHY15459S
Encompass_Preferred
Cabazitaxel-d6 is the deuterium labeled Cabazitaxel, a semi-synthetic derivative of the natural taxoid 10-deacetylbaccatin III with potential antineoplastic activity. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs. This product appears as a white to off-white solid and is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-65368