Learn More
Cayman Chemical CabazItaxel-d9 1mg
SDP
Supplier:  Cayman Chemical 284881 MG
Encompass_Preferred
An internal standard for the quantification of cabazitaxel by GC- or LC-MS
Catalog No. 50-293-9850