Learn More
Cayman Chemical CabazItaxel-d9 5mg
SDP
Supplier:  Cayman Chemical 284885 MG
Encompass_Preferred
An internal standard for the quantification of cabazitaxel by GC- or LC-MS
Catalog No. 50-293-8912