Learn More
Hach Company Hach Cable Assembly for Process Analyzer Air Compressor (MPN 6784500)
Supplier:  Hach Company 6784500
Encompass
This cable assembly provides the electrical and mechanical connection for an air compressor used with process analyzers. It is intended as a replacement or spare assembly to restore compressor function in analyzer systems.
  • Connects air compressor to process analyzer compressor system.
  • Intended for use as a replacement or spare assembly.
  • Provides electrical and mechanical connection required for compressor operation.
  • Available through manufacturer sales channels; contact manufacturer for compatibility and pricing.
Catalog No. NC3303039