Learn More
HARVARD BIOSCIENCE INC CABLE ASSEMBLY CC 28
Supplier:  HARVARD BIOSCIENCE INC 640106
Encompass_Preferred
Total order minimum requirement of $500. If only item on order, must order QTY of 2.Cable Assembly, For heater controllers, the CC-28 cable assembly has two thermistors at the platform end, The control thermistor (T1) which attaches to the platform and a bead thermistor (T2) for monitoring the bath Temperature
Catalog No. NC9975713