Learn More
Grainger Cable Stripper 24 AWG 6-1/4 In
Supplier:  Grainger 14G834
Encompass_Preferred
The Siemens Cable Stripper is designed for stripping cables and wires from 24 AWG to 14 AWG. It features precision ground stripping holes for clean, accurate strips and has an ergonomic, non-slip grip for comfort and control. The tool is constructed from hardened steel for durability. Overall, this is a high-quality manual cable stripper suitable for a range of wire gauges. It provides precise, consistent stripping and is built to withstand repeated use. The ergonomic design makes it comfortable for extended use.
Catalog No. 50-275-3644