Learn More
Cole-Parmer 40 Pound Ladder-Design Nylon Cable/Zip Ties, 11" L, White; 1000/Pk
Supplier:  Cole-Parmer 0683025
Encompass_Preferred
These ties are injection-molded of 6/6 nylon— won’t corrode or stretch. Durable locking mechanism holds securely even when used to bundle vibrating or flexing materials. Ties continue to hold tension even in oily spray or wet environments. Ties are self-extinguishing as per UL 94 Flam. class: V-2. Ties withstand temperatures to 185degF (85degC). All ties remain flexible to –40degF (–40degC). Efficient range of ties may be higher or lower in your specific application. Melting temperature is 495degF (257degC).
Catalog No. NC9668213