Learn More
HARVARD BIOSCIENCE INC CABLE W/ BEAD THERMISTOR
Supplier:  HARVARD BIOSCIENCE INC 640107
Encompass_Preferred
Total order minimum requirement of $500. If only item on order, must order QTY of 5.Cable with bead thermistor, BNC end, included with most inline solution heaters. Thermistor can be replaced by TS-70B. Thermistor dimensions are 0.85 x 4.6 mm.
Catalog No. NC0470041