Learn More
Medchemexpress LLC Cabozantinib | Purity: 99.96 | Mol wt: 501.51 | 849217-68-1 | 200mg
SDP
Supplier:  Medchemexpress LLC HY13016200MG
Encompass_Preferred
Cabozantinib | Purity: 99.96 | Mol wt: 501.51 | 849217-68-1 | 200mg
Catalog No. NC2037934