Learn More
Medchemexpress LLC Cabozantinib-d6 | 1802168-46-2 | 98.1% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY13016S1MG
Encompass_Preferred
Cabozantinib-d6 is a deuterium-labeled derivative of Cabozantinib, a potent inhibitor targeting multiple receptor tyrosine kinases (RTKs) including VEGFR2, c-Met, Kit, Axl, and Flt3. This compound is designed for research applications, serving as a valuable tracer or internal standard for precise quantitative analysis methods such as NMR, GC-MS, and LC-MS.
  • Deuterium-labeled compound
  • Potent inhibitor of multiple receptor tyrosine kinases
  • Inhibits VEGFR2, c-Met, Kit, Axl, and Flt3
  • Useful as a tracer in research
  • Functions as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, and LC-MS techniques
  • Intended for research use only
Catalog No. 50-003-38566