Learn More
Medchemexpress LLC Cabozantinib-d6 | 1802168-46-2 | 98.1% | C28H18D6FN3O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13016S10MG
Encompass_Preferred
Cabozantinib-d6 is a deuterium-labeled analog of cabozantinib supplied as a 10 mg reference standard for use as a stable isotope internal standard in LC-MS quantitative and pharmacokinetic assays. It supports accurate quantification and method validation in analytical laboratories.
Catalog No. 50-002-58842