Learn More
TARGETMOL CHEMICALS INC Cabozantinib hydrochloride 2
SDP
Supplier:  TARGETMOL CHEMICALS INC T516425MG
Encompass_Preferred
Also available in 1 mg, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Cabozantinib hydrochloride (XL184) is a potent pan-tyrosine kinases inhibitor that inhibits VEGFR2, c-Met, Kit, Axl, and Flt4 (IC50s 0.035, 1.3, 4.6, 7 and 6 nM). Purity 97.09%
Catalog No. 50-287-9784