Learn More
AdipoGen Cabozantinib S-malate (100 mg)
SDP
Supplier:  AdipoGen CDXC0698M100
Encompass_Preferred
Chemical. CAS: 1140909-48-3. Formula: C32H30FN3O10. MW: 635.59. The s-malate salt form of Cabozantinib is very potent orally available inhibitor of multiple receptor tyrosine kinases (RTK), specifically MET and VEGFR2. It also inhibits KIT, FLT3, Tie-2, RET and AXL. Cabozantinib shows dose-dependent inhibition of tumor growth and tumor regression, associated with disruption of the tumor vasculature, angiogenesis and extensive tumor cell apoptosis.
Catalog No. 50-205-4354