Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cabozantinib S-malate | 1140909-48-3 | 99.9% | 500 MG
SDP

Supplier:  Medchemexpress LLC HY12044500MG

Encompass_Preferred

Cabozantinib S-malate is the (S)-malate salt of cabozantinib, a multi-receptor tyrosine kinase inhibitor used in research to probe VEGFR2, c-Met, Kit, Axl, and Flt3 signalling. It is supplied as a high-purity analytical standard in solid form (for example, 500 MG) and as a 10 mM solution in DMSO for convenient use.

  • High purity, 99.9%.
  • Molecular formula C32H30FN3O10, molecular weight 635.59 g/mol.
  • Research applications include kinase inhibition and signaling pathway studies.
  • Available as a solid and as a 10 mM solution in DMSO for reconstitution or direct use.
  • CAS number 1140909-48-3 for unambiguous identification.

Catalog No. 50-002-33260


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.