Learn More
Medchemexpress LLC HY-100611 5mg Medchemexpress, CaCCinh-A01 CAS:407587-33-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1006115MG
Encompass_Preferred
Medchemexpress, HY-100611 5mg CaCCinh-A01 CAS:407587-33-1 CaCCinh-A01 is an inhibitor of both TMEM16A and calcium-activated chloride channel ( CaCC ) with IC 50 s of 2.1 and 10 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1900