Learn More
Selleck Chemical LLC CaCCinh-A01 S0125-25MG
SDP
Supplier:  Selleck Chemical LLC S012525MG
Encompass_Preferred
CaCCinh-A01 (TMEM16 Blocker I) is a blocker of TMEM16A with IC50 of 2 1 M CaCCinh-A01 is also an inhibitor of calcium-activated chloride channel (CaCC) with IC50 of 10 M
Catalog No. 50-000-39726