Learn More
TARGETMOL CHEMICALS INC CADAZOLID 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T402010MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cadazolid (ACT-179811) is an oxazolidinone-type antibiotic with activity against gram-positive bacteria including Clostridium difficile. purity: 99%
Catalog No. 50-257-6827