Learn More
Medchemexpress LLC Cadherin-1/CD324 Hu 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP76975UG
Encompass_Preferred
Cadherin-1/CD324 Protein Human (HEK293 His) is a recombinant human E-cadherin produced in HEK293 cells with His tag Cadherin-1/CD324/E-cadherin is one of the most important molecules in cell-cell adhesion in epithelial tissues[1]
Catalog No. 50-001-85869