Learn More
Medchemexpress LLC Cadherin-17 protein, cynomolgus (biotinylated, HEK293, His-Avi) | ≥ 90% | Predicted molecular mass is 88.1 kDa | 200 UG
SDP
Supplier:  Medchemexpress LLC 200UGHYP705186
Encompass_Preferred
The biological activity of this Cadherin-17 protein is measured by its binding ability in a functional ELISA. When immobilized at 1 μg/mL on a streptavidin precoated plate, the Biotinylated Cynomolgus Cadherin-17 Protein, His, Avitag can bind Anti-CDH17 antibody (300-350) with a linear range of 2.924-38.1 ng/mL.
  • The protein is expressed in HEK293 cells.
  • It is biotinylated with a C-Avi and C-His tag.
  • The product is supplied as a lyophilized powder.
  • It is formulated from a 0.22 μm filtered solution in 20 mM Tris, 150 mM NaCl, pH 8.0, with trehalose as a protectant.
  • It can be stored at -20°C for 2 years from the date of receipt.
  • After reconstitution, it remains stable at 4°C for 1 week or at -20°C for longer with a carrier protein.
  • Suitable for use in functional ELISA to assess binding capabilities.
  • Provides a standardized method for evaluating binding affinity to antibodies.
Catalog No. 50-003-64058