Learn More
Medchemexpress LLC δ-Cadinene | 483-76-1 | 204.35 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN10544100MG
Encompass_Preferred
δ-Cadinene is a sesquiterpene found in essential oils with significant biological activities. It demonstrates antiproliferative and pro-apoptotic effects on human ovarian cancer (OVCAR-3) cells, and also possesses trichomonacidal, antimicrobial, antifungal, and anticancer properties.
  • Possesses trichomonacidal properties.
  • Shows antimicrobial and antifungal activity.
  • Exhibits anticancer effects.
  • Inhibits OVCAR-3 cell growth in a concentration-dependent manner.
  • Induces both early and late apoptosis in OVCAR-3 cells.
  • Leads to sub-G1-phase growth arrest in OVCAR-3 cells.
Catalog No. 50-004-16566