Learn More
Strem, An Ascensus Company CAS# 543-90-8. 5g. Cadmium acetate, anhydrous (99.999%-Cd) PURATREM. MFCD00012449
SDP
Supplier:  Strem, An Ascensus Company 4801005G
Encompass
CAS# 543-90-8. 5g. Cadmium acetate, anhydrous (99.999%-Cd) PURATREM. MFCD00012449. Molecular Weight: 230.50. Molecular Formula: Cd(OOCCH3)2. Color/form: white pwdr. Strem# 48-0100. http://www.strem.com/catalog/v/48-0100/
Catalog No. 50-901-12543