Learn More
Strem, An Ascensus Company CAS# 10108-64-2. 10g. Cadmium chloride, anhydrous (99.995%-Cd) PURATREM. MFCD00010916
SDP
Supplier:  Strem, An Ascensus Company 48150010G
Encompass
CAS# 10108-64-2. 10g. Cadmium chloride, anhydrous (99.995%-Cd) PURATREM. MFCD00010916. Molecular Weight: 183.32. Molecular Formula: CdCl2. Color/form: white pwdr. Strem# 48-1500. http://www.strem.com/catalog/v/48-1500/
Catalog No. 50-901-12550