Learn More
Strem, An Ascensus Company CAS# 1306-19-0. 25g. Cadmium oxide (99.999%-Cd) PURATREM. MFCD00010921
SDP
Supplier:  Strem, An Ascensus Company 48080025G
Encompass
CAS# 1306-19-0. 25g. Cadmium oxide (99.999%-Cd) PURATREM. MFCD00010921. Molecular Weight: 128.40. Molecular Formula: CdO. Color/form: red to brown pwdr. Strem# 48-0800. http://www.strem.com/catalog/v/48-0800/
Catalog No. 50-901-12547