Learn More
Selleck Chemical LLC Caerulein FI-6934-1mg
SDP
Supplier:  Selleck Chemical LLC S96901MG
Encompass_Preferred
Caerulein (Ceruletide, Cerulein, FI-6934), a cholecystokinin (CCK) receptor agonist, is a safe and effective cholecystokinetic agent and small bowel and exocrine pancreatic stimulant.
Catalog No. 50-238-9955