Learn More
Medchemexpress LLC Caerulomycin A 10mg | 21802-37-9 | 229.24 g/mol | C12H11N3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11449510MG
Encompass_Preferred
Caerulomycin A is a small-molecule antibiotic research reagent with reported antifungal and immunomodulatory activity. Supplied as a white to off-white solid, it is intended for in vitro and in vivo laboratory studies; reported purity is 98.0% and molecular weight is 229.24 g/mol.
  • High reported purity suitable for research applications.
  • Supplied in mg-scale quantities appropriate for screening and pharmacology studies.
  • Soluble in DMSO with specified in vitro and in vivo vehicle formulations.
  • White to off-white solid, straightforward to handle and store.
  • Manufacturer-provided solubility and dosing protocols available for in vivo use.
Catalog No. 50-002-29724