Learn More
TARGETMOL CHEMICALS INC Caerulomycin A 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T148541MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Caerulomycin A (Caerulomycin) (Cerulomycin; Caerulomycin) is an antifungal compound. Which induces generation of T cells, enhances TGF-beta-Smad3 protein signaling via suppressing interferon-gamma-induced STAT1 signaling. Antifungal and antibiotic activity, and used in autoimmune diseases. Purity 97.51%
Catalog No. 50-287-6401