Learn More
TARGETMOL CHEMICALS INC CAFESTOL 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T572725MG
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. CAFESTOL is a ERK inhibitor for AP-1-targeted activity against PGE2 production and the mRNA expression of cyclooxygenase (COX)-2 in LPS-activated RAW264.7 cells. Cafestol has strong inhibitory activity on PGE2 production by suppressing the NF-kB activation pathway. Purity 99.84%
Catalog No. 50-287-6978