Learn More
Medchemexpress LLC Cafestol | 469-83-0 | 20 MG
SDP
Supplier:  Medchemexpress LLC HYN625720MG
Encompass_Preferred
Cafestol is an orally active diterpenoid and an inhibitor of ERK2. It exhibits various biological activities including anti-inflammatory, anti-angiogenic, and anti-diabetic effects. This compound also induces tumor cell apoptosis and autophagy, making it a valuable tool for research, particularly in cancer studies.
  • Inhibitor of ERK2
  • Exhibits anti-inflammatory activity
  • Shows anti-angiogenic activity
  • Demonstrates anti-diabetic effects
  • Induces tumor cell apoptosis
  • Induces autophagy
  • Targets COX-2 and NF-κB pathways
  • Inhibits AP-1 activation
Catalog No. 50-004-18750