Learn More
Cayman Chemical CaffeIc AcId 5g
SDP
Supplier:  Cayman Chemical 706025 G
Encompass_Preferred
A polyphenol with diverse biological activities; inhibits KDM6A and KDM4C (IC50s = 5.5 and 13.7 µM, respectively); inhibits 12-LO and 5-LO (IC50s = 5.1 and 72 µM, respectively); inhibits MMP-9 and MMP-2 (IC50s = 8 and 12 µM, respectively); inhibits PMA-induced increases in migration and invasion in HepG2 cells at 100 UG/ml; decreases tumor growth and the number of liver metastases in a HepG2 mouse xenograft model at 5 mg/kg three times per week
Catalog No. 50-289-6203