Learn More
Selleck Chemical LLC Caffeic Acid Phenethyl Ester 50mg 104594-70-9 CAPE, Phenylethyl Caffeate
SDP
Supplier:  Selleck Chemical LLC S741450MG
Encompass_Preferred
Caffeic acid phenethyl ester (CAPE, Phenylethyl Caffeate) is a potent and specific inhibitor of NF-?B activation, and also displays antioxidant, immunomodulatory and antiinflammatory activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1344589