Learn More
Abraxis Caffeine, ELISA, 96-test
SDP
Supplier:  Abraxis 515575
Encompass
The Caffeine ELISA is an immunoassay for the quantitative and sensitive detection of Caffeine. This test is suitable for the quantitative and/or qualitative screening of Caffeine in water, urine, and saliva samples. If necessary, positive samples can be confirmed by HPLC, GC/MS, or other conventional methods. Note: This assay is intended For Research Use Only. The test is a direct competitive ELISA which is based on the recognition of Caffeine by specific antibodies.
Catalog No. NC1710833