Learn More
Abcam Caged MK 801, NMDA receptor open-channel blocker, 50MG
Supplier:  Abcam AB14643150MG
Encompass_Preferred
MW 460.49 g/mol, Purity >99%. Caged, potent, selective, non-competitive, irreversible NMDA receptor open-channel blocker. UV-active MK 801 (ab144485, ab120027, ab120028) analog. Shows anticonvulsant effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4618