Learn More
Genetex Inc Calbindin Monoclonal Antibody [HMV326] HistoMAX™ (GTX640558) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX640558
Encompass_Preferred
The Calbindin antibody [HMV326] HistoMAX™ (GTX640558) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human cerebellum cortex tissue (molecular layer, Purkinje cell layer, and granule cell layer) using GTX640558 Calbindin antibody [HMV326] HistoMAX. Strong Calbindin staining of Purkinje cells and of associated axonal fibres.
Catalog No. NC3669338