Learn More
Genetex Inc Calbindin Monoclonal Antibody [MSVA-471M] HistoMAX™ (GTX04454) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04454
Encompass_Preferred
The Calbindin antibody [MSVA-471M] HistoMAX™ (GTX04454) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human cerebellum tissue (molecular layer, Purkinje cell layer, granule cell layer, and white matter) using GTX04454 Calbindin antibody [MSVA-471M] HistoMAX. A strong Calbindin staining of Purkinje cells and of associated axonal fibres is seen in the cerebrum.
Catalog No. NC3667729