Learn More
Medchemexpress LLC Calcein tetraethyl ester | 1170856-93-5 | MFCD14586332 | 99.5% | 734.75 | C38H42N2O13 | 10 MG

Supplier: Medchemexpress LLC HYD083010MG
Calcein (tetraethyl ester) is a membrane-permeant fluorescent dye used to label live cells in viability assays, cell tracing, and fluorescence microscopy. The esterified form crosses cell membranes and is hydrolyzed intracellularly to fluorescent calcein, producing strong green fluorescence for imaging and quantitative measurements.
- Membrane-permeant fluorescent dye for live-cell labeling
- Intracellular conversion yields bright green fluorescence
- High purity suitable for research use (99.5%)
- Molecular weight 734.75 g/mol
- CAS number 1170856-93-5 for identification
- Available in small quantities for laboratory assays
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.