Learn More
TARGETMOL CHEMICALS INC CALCIFEDIOL 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T643010MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Calcifediol (U 32070E)(Ki of 3.9 uM) a major circulating metabolite of vitamin D3 works as a competitive inhibitor. It (ED50=2 nM)also inhibits PTH secretion and mRNA. purity: 99%
Catalog No. 50-257-9380