Learn More
Medchemexpress LLC Calcifediol-d3 | 140710-94-7 | 99.0% | 403.66 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY32351S5MG
Encompass_Preferred
Calcifediol-d3 is a deuterium labeled Calcifediol. Calcifediol, is an effective VDR ligand and VD supplement. Calcifediol is a prohormone of the vitamin D endocrine system (VDES) and is hydroxylated in the liver to produce the active form, calcitriol. Calcifediol can rapidly increase serum VD levels.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-04749