Learn More
Medchemexpress LLC HY-N6687 1mg Medchemexpress, Calcimycin CAS:52665-69-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN66871MG
Encompass_Preferred
Medchemexpress, HY-N6687 1mg Calcimycin CAS:52665-69-7 Calcimycin (A23187) is an antibiotic and a unique divalent cation ionophore (like calcium and magnesium). It induces Ca 2+ -dependent cell death by increasing intracellular calcium concentration. Calcimycin inhibits the growth of Gram-positive bacteria and some fungi. Calcimycin also inhibits the activity of ATPase and uncouples oxidative phosphorylation of mammalian cells. It induces apoptosis [1] [2] [3] [4] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3968