Learn More
Medchemexpress LLC Calcineurin A (PPP3CA), human recombinant | 90.0% | 50 UG

Supplier: Medchemexpress LLC HYP7675950UG
Recombinant human Calcineurin A (PPP3CA), expressed in Sf9 insect cells with a His tag, supplied as 50 μg of purified protein (~60 kDa). Intended for enzymatic assays, biochemical characterization, and antibody generation; store at -20°C.
- Recombinant human catalytic subunit of calcineurin.
- Expressed in Sf9 insect cells for eukaryotic folding.
- His-tagged for affinity purification and detection.
- Approximate molecular weight 60 kDa.
- Purity 90.0% for biochemical applications.
- Supplied as 50 μg with recommended storage at -20°C.
- Suitable for enzyme activity assays and antibody production.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.